Thermo Scientific Chemicals

Aluminum phosphate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010628.06
5 g, Each
Thermo Scientific Chemicals

Aluminum phosphate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010628.06
5 g, Each
Quantity
Catalog number: 010628.06
also known as 010628-06
Price (USD)
Price: 94.20
Online price: 80.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7784-30-7
IUPAC Name
aluminium(3+) phosphate
Molecular Formula
AlO4P
InChI Key
ILRRQNADMUWWFW-UHFFFAOYSA-K
SMILES
[Al+3].[O-]P([O-])([O-])=O
Total Metal Impurities
0.01% max.

Description

Aluminum phosphate is used in the production of glass, mixture of ceramic, waterproofing concrete and dental cements, in cosmetics as a emollient, flame retardant, catalyst in organic synthesis, fireproofing textile, pharmaceuticals and dyes. It also used in many adhesives, binders and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text