Thermo Scientific Chemicals

Barium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010647.14
25 g, Each
Thermo Scientific Chemicals

Barium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010647.14
25 g, Each
Quantity
Catalog number: 010647.14
also known as 010647-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
7787-32-8
IUPAC Name
barium(2+) difluoride
Molecular Formula
BaF2
InChI Key
OYLGJCQECKOTOL-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ba++]
Appearance (Color)
Colorless or white
Form
Crystals or powder or crystalline powder
Total Metal Impurities
≤0.01% (≤100ppm)

Description

Barium fluoride is used in windows for thermal imaging inspection application in electric power facilities. Barium fluoride can be used to make optical components like lenses, as it is transparent in UV (ultraviolet) and IR (infrared) regions. It is used in windows for IR fuel oil analysis. Barium fluoride is one of the fastest scintillators for the detection of X-rays, and gamma rays. It has been found to be useful for the detection of gamma photons in PET (positron emission tomography), high energy neutrons, alpha and beta particles. Its resistant to high energy radiation is higher than all other optical fluorides. It is employed in metallurgy for refining aluminum, production of welding agents, as a preopacifying agent and in glazing frits production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text