Barium nitrate, Puratronic™, 99.999% (metals basis)
Barium nitrate, Puratronic™, 99.999% (metals basis)
Barium nitrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Barium nitrate, Puratronic™, 99.999% (metals basis)

CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
產品號碼 010648.36
亦稱為 010648-36
價格 (TWD)
-
Quantity:
500 g
Request bulk or custom format
化學識別
CAS10022-31-8
IUPAC Namebarium(2+) dinitrate
Molecular FormulaBaN2O6
InChI KeyIWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
FormCrystals or powder
Assay from Suppliers CofA±2.5 % of Theoretical value (Ba)
Total Metal Impurities≤0.001% (10 ppm)
Barium nitrate is used by the oil and gas industries to make drilling muds. It is used to make  paints, bricks, tiles, glass, rubber, and other barium compounds. It also used in is used in signal flares and water softener. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only