Thermo Scientific Chemicals

Cadmium carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010660.18
50 g, Each
Thermo Scientific Chemicals

Cadmium carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010660.18
50 g, Each
Quantity
Catalog number: 010660.18
also known as 010660-18
Price (USD)

Chemical Identifiers

CAS
513-78-0
IUPAC Name
cadmium(2+) carbonate
Molecular Formula
CCdO3
InChI Key
GKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES
[Cd++].[O-]C([O-])=O
Total Metal Impurities
0.002% max.

Description

Cadmium carbonate is the raw material for the production of pigments for ceramic and frits and for production of cadmium salt. Used as reagents. It is used for intermediates of manufacturing polyester, glass pigment flux, organic catalysts, plastic plasticizer, stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text