Thermo Scientific Chemicals

Calcium chloride hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

Catalog number: 010680.09
10 g, Each
Thermo Scientific Chemicals

Calcium chloride hydrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals

Catalog number: 010680.09
10 g, Each
Quantity
Catalog number: 010680.09
also known as 010680-09
Price (USD)
Price: 136.00
Online price: 115.65
Your price:
Quantity
-

Chemical Identifiers

CAS
22691-02-7
Total Metal Impurities
0.004% max.

Description

It is used as a food additive, as an electrolyte, it is used as a buffer in pools, an additive in plastics and in fire extinguishers. It is used as a fabric softener, dust control on roads. Calcium chloride is a soluble compound that can serve as a source of calcium ions in a solution. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text