Thermo Scientific Chemicals

Ammonium hexachloroiridate(IV), Premion™, 99.994% (metals basis), Ir 41% min, Thermo Scientific Chemicals

Catalog number: 010712.77
0.1 g, Each
Thermo Scientific Chemicals

Ammonium hexachloroiridate(IV), Premion™, 99.994% (metals basis), Ir 41% min, Thermo Scientific Chemicals

Catalog number: 010712.77
0.1 g, Each
Quantity
Catalog number: 010712.77
also known as 010712-77
Price (USD)

Chemical Identifiers

CAS
16940-92-4
IUPAC Name
diammonium hexachloroiridiumtetrakis(ylium)
Molecular Formula
Cl6H8IrN2
InChI Key
QOPXOJZBUJGPAG-UHFFFAOYSA-J
SMILES
[NH4+].[NH4+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl
Total Metal Impurities
0.006% max.
Assay (unspecified)
Ir 41% min

Description

It is a key intermediate in the isolation of iridium from ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text