Thermo Scientific Chemicals

Lithium iodide hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010738.14
25 g, Each
Thermo Scientific Chemicals

Lithium iodide hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010738.14
25 g, Each
Quantity
Catalog number: 010738.14
also known as 010738-14
Price (USD)
Price: 253.00
Online price: 215.65
Your price:
Quantity
-

Chemical Identifiers

CAS
85017-80-7
IUPAC Name
lithium(1+) iodide
Molecular Formula
ILi
InChI Key
HSZCZNFXUDYRKD-UHFFFAOYSA-M
SMILES
[Li+].[I-]
Total Metal Impurities
0.005% max.

Description

Lithium iodide hydrate is used as a reagent for ester cleavage and decarboxylation and C-C bond forming reactions. It is also used as source of nucleophilic iodide, as a mild lewis acid, epoxide opening and as an additive for organometallic-mediated transformations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text