Lithium niobium oxide, Puratronic™, 99.998% (metals basis excluding Ta), Ta <50ppm
Lithium niobium oxide, Puratronic™, 99.998% (metals basis excluding Ta), Ta <50ppm
Lithium niobium oxide, Puratronic™, 99.998% (metals basis excluding Ta), Ta <50ppm
Thermo Scientific Chemicals

Lithium niobium oxide, Puratronic™, 99.998% (metals basis excluding Ta), Ta <50ppm

CAS: 12031-63-9 | LiNbO3 | 147.84 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
產品號碼 010741.14
亦稱為 010741-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS12031-63-9
IUPAC Namelithium(1+) niobiumoylolate
Molecular FormulaLiNbO3
InChI KeyGQYHUHYESMUTHG-UHFFFAOYSA-N
SMILES[Li+].[O-][Nb](=O)=O
檢視更多
規格Specification Sheet規格表
Impurity content50ppm Ta max
Total Metal Impurities excluding Ta0.002% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only