Thermo Scientific Chemicals

Hafnium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Catalog number: 010793.FP
80 x 250 mm, Each
Thermo Scientific Chemicals

Hafnium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Catalog number: 010793.FP
80 x 250 mm, Each
Quantity

Chemical Identifiers

CAS
7440-58-6
IUPAC Name
hafnium
Molecular Formula
Hf
InChI Key
VBJZVLUMGGDVMO-UHFFFAOYSA-N
SMILES
[Hf]
Trace Metal
Total metal impurities less than 0.5% excluding Zr, Zr nominal 3%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text