Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010795.22
100 g, Each
Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010795.22
100 g, Each
Quantity
Catalog number: 010795.22
also known as 010795-22
Price (USD)
Price: 425.00
Online price: 368.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16674-78-5
IUPAC Name
magnesium(2+) diacetate tetrahydrate
Molecular Formula
C4H14MgO8
InChI Key
XKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILES
O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Total Metal Impurities
0.003% max.

Description

Used as a source of magnesium in biological reactions, and as a source of acetate ion in chemical transformations. Acts as an emission control agent in coal combustion process to reduce acid rain. It is involved in glass formation process via the thermal decomposition of solids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text