Thermo Scientific Chemicals

Mercury(II) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010808.14
25 g, Each
Thermo Scientific Chemicals

Mercury(II) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010808.14
25 g, Each
Quantity
Catalog number: 010808.14
also known as 010808-14
Price (USD)

Chemical Identifiers

CAS
7487-94-7
IUPAC Name
mercury(2+) dichloride
Molecular Formula
Cl2Hg
InChI Key
LWJROJCJINYWOX-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Hg++]
Total Metal Impurities
0.001% max.

Description

Mercuric chloride is used as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride It is used as embalming chemical, photographic intensifier. It is also used as a treatment for syphilis and in very dilute solution as an antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text