Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)
Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)
Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)
Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)

CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 010813.06
亦稱為 010813-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS6018-89-9
IUPAC Namenickel(2+) diacetate tetrahydrate
Molecular FormulaC4H14NiO8
InChI KeyOINIXPNQKAZCRL-UHFFFAOYSA-L
SMILESO.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
檢視更多
規格Specification Sheet規格表
Total Metal Impurities0.001% max.
It is used in electroplating, anodize coatings; textile dyeing mordant; Catalyst. Also, used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only