Thermo Scientific Chemicals

Potassium nitrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010844.18
50 g, Each
Thermo Scientific Chemicals

Potassium nitrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010844.18
50 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 010844.18
also known as 010844-18
Price (USD)
Price: 151.00
Your price:
Quantity
-

Chemical Identifiers

CAS
7757-79-1
IUPAC Name
potassium nitrate
Molecular Formula
KNO3
InChI Key
FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES
[K+].[O-][N+]([O-])=O
Appearance (Color)
White
Form
Crystalline powder and/or granules
Total Metal Impurities
≤0.005%

Description

Potassium nitrate has a vast variety of applications. Appropriate preparations of potassium nitrate find use in fertilizers, food preservatives, tooth paste, rocket propellants and fireworks. In addition, it is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems. It forms phase change material with silica and alumina nanoparticles. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text