Thermo Scientific Chemicals

Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010863.18
50 g, Each
Thermo Scientific Chemicals

Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010863.18
50 g, Each
Quantity
Catalog number: 010863.18
also known as 010863-18
Price (USD)
Price: 344.00
Online price: 292.65
Your price:
Quantity
-

Chemical Identifiers

CAS
144-55-8
IUPAC Name
sodium hydrogen carbonate
Molecular Formula
CHNaO3
InChI Key
UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES
[Na+].OC([O-])=O
Total Metal Impurities
0.002% max.

Description

Sodium hydrogen carbonate is used in a process for removing paint and corrosion called sodablasting; the process is particularly suitable for cleaning aluminum panels which can be distorted by other types of abrasive. It can be administered to pools, spas, and garden ponds to raise pH levels. Also can be used as a fire extinguisher. Its primary use is in cooking (baking), as a leavening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text