Thermo Scientific Chemicals

Sodium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010873.06
5 g, Each
Thermo Scientific Chemicals

Sodium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010873.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 010873.06
also known as 010873-06
Price (USD)
Price: 78.70
Your price:
Quantity
-

Chemical Identifiers

CAS
7631-99-4
IUPAC Name
sodium nitrate
Molecular Formula
NNaO3
InChI Key
VWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES
[Na+].[O-][N+]([O-])=O
Appearance (Color)
White
Form
Crystalline powder
Assay (special)
?2.5 % of Theoretical value (Na)
Total Metal Impurities
≤0.001% (10 ppm)

Description

Sodium nitrate is used in the production of fertilizers, nitric acid, pyrotechnics, smoke-bombs, glass and pottery enamels. In combination with boron trifluoride it forms an efficient reagent for nitration of aromatic compounds. Adsorption on alumina provides an environmentally benign aromatic nitrating agent. Further it finds use as food preservative and as a solid rocket propellant. It is also used as an electrolyte in a salt bridge, and as thermal storage medium in power generation systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text