Tellurium(IV) oxide, Puratronic, 99.999% (metals basis)
Tellurium(IV) oxide, Puratronic, 99.999% (metals basis)
Tellurium(IV) oxide, Puratronic, 99.999% (metals basis)
Thermo Scientific Chemicals

Tellurium(IV) oxide, Puratronic, 99.999% (metals basis)

CAS: 7446-07-3 | O2Te | 159.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 010882.14
also known as 010882-14
Price (USD)
161.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
161.00
Each
Chemical Identifiers
CAS7446-07-3
IUPAC Name(oxo-λ⁴-tellanylidene)oxidane
Molecular FormulaO2Te
InChI KeyLAJZODKXOMJMPK-UHFFFAOYSA-N
SMILESO=[Te]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder
Total Metal Impurities≤0.001%
It is also used in devices which can convert light into sound (acousto-optic material). Glasses made of tellurium oxide have high refractive indices and transmit into mid-IR region. It is used to make glasses with special properties. It is useful in optical waveguides and optical fiber amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only