Thermo Scientific Chemicals

Tellurium(IV) oxide, Puratronic, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010882.36
500 g, Each
Thermo Scientific Chemicals

Tellurium(IV) oxide, Puratronic, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010882.36
500 g, Each
Quantity
Catalog number: 010882.36
also known as 010882-36
Price (USD)
Price: 1,457.00
Online price: 1,238.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7446-07-3
IUPAC Name
(oxo-λ⁴-tellanylidene)oxidane
Molecular Formula
O2Te
InChI Key
LAJZODKXOMJMPK-UHFFFAOYSA-N
SMILES
O=[Te]=O
Appearance (Color)
White to pale yellow
Form
Powder
Total Metal Impurities
≤0.001%

Description

It is also used in devices which can convert light into sound (acousto-optic material). Glasses made of tellurium oxide have high refractive indices and transmit into mid-IR region. It is used to make glasses with special properties. It is useful in optical waveguides and optical fiber amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text