Carbon, activated, 2mm & down
Carbon, activated, 2mm & down
Carbon, activated, 2mm & down
Thermo Scientific Chemicals

Carbon, activated, 2mm & down

For liquid purification | CAS: 1333-86-4 | C | 12.01 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
250 g
1 kg
產品號碼 010926.30
亦稱為 010926-30
價格 (TWD)
-
Quantity:
250 g
Request bulk or custom format
規格
CAS1333-86-4
Chemical Name or MaterialCarbon
EINECS Number231-153-3
Health Hazard 1H500
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
檢視更多
For liquid purification

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only