Iron wire, 0.5mm (0.02in) dia, Puratronic™, 99.995% (metals basis)
Iron wire, 0.5mm (0.02in) dia, Puratronic™, 99.995% (metals basis)
Iron wire, 0.5mm (0.02in) dia, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Iron wire, 0.5mm (0.02in) dia, Puratronic™, 99.995% (metals basis)

CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 010936.G1
also known as 010936-G1
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
178.65
Online Exclusive
198.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.5mm (0.02in) diameter.
Assay from Supplier's CofA≥99.995% (metals basis)
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only