Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis)
Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis)
Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis)
Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis)
Thermo Scientific Chemicals

Platinum wire, 1.0mm (0.04in) dia, Premion™, 99.997% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
2 cm
10 cm
Catalog number 010956.BW
also known as 010956-BW
Price (USD)
1,931.65
Special offer
Online exclusive
2,272.00
Save 340.35 (15%)
Each
Add to cart
Quantity:
50 cm
Request bulk or custom format
Price (USD)
1,931.65
Special offer
Online exclusive
2,272.00
Save 340.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.003% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only