Thermo Scientific Chemicals

Potassium carbonate sesquihydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals

Catalog number: 010981.36
500 g, Each
Thermo Scientific Chemicals

Potassium carbonate sesquihydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals

Catalog number: 010981.36
500 g, Each
Quantity
Catalog number: 010981.36
also known as 010981-36
Price (USD)
Price: 60.00
Online price: 51.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6381-79-9
IUPAC Name
tetrapotassium trihydrate dicarbonate
Molecular Formula
C2H6K4O9
InChI Key
WWVGGBSGYNQKCY-UHFFFAOYSA-J
SMILES
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
Heavy Metals
(ICP-OES) 5ppm
Impurity content
Ca 0.005%
Impurity content
Cl 0.003%
Impurity content
SiO₂ 0.005%
Impurity content
Sulfur compounds (as SO₄) 0.004%

Description

Potassium carbonate sesquihydrate is used for manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts and as an analytical lab reagent. It is also useful as a reagent for removing carbon dioxide from ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text