Thermo Scientific Chemicals

Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals

250 mL, Each
Thermo Scientific Chemicals

Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals

250 mL, Each
Quantity

Chemical Identifiers

CAS
7697-37-2
IUPAC Name
nitric acid
Molecular Formula
HNO3
InChI Key
GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES
O[N+]([O-])=O
Appearance (Color)
Clear colorless
Form
Liquid
Assay from Supplier's CofA
65.0-70.0%
Total Metal Impurities
≤0.0001%

Description

Nitric acid, 65-70% is used in as a nitrating or an oxidizing agent in organic synthesis. It is industrially used for the manufacturing of fertilizers. Further, it finds use in manufacture of explosives, nylon precursors, specialty organic compounds and drug intermediates.It is also used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple. It is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only