Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals
Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals
Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitric acid, 65-70%, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
變更視圖buttonViewtableView
Quantity:
25 mL
250 mL
500 mL
產品號碼 010984.AP
價格 (TWD)
-
Quantity:
500 mL
Request bulk or custom format
化學識別
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless
Assay from Supplier's CofA65.0-70.0%
Total Metal Impurities≤0.0001%
FormLiquid

Nitric acid, 65-70% is used in as a nitrating or an oxidizing agent in organic synthesis. It is industrially used for the manufacturing of fertilizers. Further, it finds use in manufacture of explosives, nylon precursors, specialty organic compounds and drug intermediates.It is also used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple. It is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only