Lithium nitrate, anhydrous, 99.98% (metals basis)
Lithium nitrate, anhydrous, 99.98% (metals basis)
Lithium nitrate, anhydrous, 99.98% (metals basis)
Thermo Scientific Chemicals

Lithium nitrate, anhydrous, 99.98% (metals basis)

CAS: 7790-69-4 | LiNO3 | 68.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
250 g
Catalog number 010985.09
also known as 010985-09
Price (EUR)
67,20
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
67,20
Each
Chemical Identifiers
CAS7790-69-4
IUPAC Namelithium(1+) nitrate
Molecular FormulaLiNO3
InChI KeyIIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES[Li+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.02% max.
Lithium nitrate, anhydrous is used in the manufacture of red-colored fireworks and flares. has been proposed as a medium to store heat collected from the sun for cooking. A Fresnel lens would be used to melt solid lithium nitrate, which would then function as a 'solar battery', allowing heat to be redistributed later by convection

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only