Orion™ Conductivity Conditioning Solution; 5 x 60 mL bottles
Actual product may vary
Orion™ Conductivity Conditioning Solution; 5 x 60 mL bottles
Orion™ Conductivity Conditioning Solution; 5 x 60 mL bottles
Thermo Scientific™

Orion™ Conductivity Conditioning Solution; 5 x 60 mL bottles

Perform virtually any electrochemistry analysis with these conductivity standards and solutions. All conductivity standards are traceable to NIST™ standard reference materials.
Have Questions?
Catalog NumberQuantity
01100160 mL
Catalog number 011001
Price (USD)
97.20
Online exclusive
108.00
Save 10.80
Case of 5
Quantity:
60 mL
Price (USD)
97.20
Online exclusive
108.00
Save 10.80
Case of 5

Perform virtually any electrochemistry analysis with Thermo Scientific™ Orion™ Conductivity Standards and Solutions. Choose from a complete selection of conductivity accessories including conductivity standards and TDS (total dissolved solids) standards. All conductivity standards are traceable to NIST™ standard reference materials.

Conductivity Standards

  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
Specifications
Boiling Point100°C
ColorLight Green
Conductivity—
DescriptionConductivity Conditioning Solution
For Use With (Equipment)Conductivity Probes (011050MD, 011050A and 011050)
PackagingBottle
Quantity60 mL
SolubilitySoluble
Solution TypeConditioning Solution
Specific Gravity980 kg/m3
Traceability to NISTYes
TypeConductivity Standard
pH11.5
Unit SizeCase of 5