Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min
Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min
Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min
Thermo Scientific Chemicals

Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min

CAS: 13454-96-1 | Cl4Pt | 336.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 011045.03
also known as 011045-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS13454-96-1
IUPAC Nametetrachloroplatinum
Molecular FormulaCl4Pt
InChI KeyFBEIPJNQGITEBL-UHFFFAOYSA-J
SMILESCl[Pt](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay (special)Pt 57.0-57.9%
Total Metal Impurities≤0.1%
FormCrystalline
Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only