Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011067.A1
Price (USD)/ Each
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
Molecular Weight (g/mol)26.98
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or flake
Total Metal Impurities≤ 0.5%
Particle Size-325 mesh (less than 45 micron)
Trace MetalsTrace metals ≤ 0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only