Copper powder, spherical, -100 mesh, 99.5% (metals basis)
Copper powder, spherical, -100 mesh, 99.5% (metals basis)
Copper powder, spherical, -100 mesh, 99.5% (metals basis)
Copper powder, spherical, -100 mesh, 99.5% (metals basis)
Thermo Scientific Chemicals

Copper powder, spherical, -100 mesh, 99.5% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 x 1 kg
Catalog number 011070.D9
also known as 011070-D9
Price (EUR)
-
Quantity:
5 x 1 kg
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, -100 mesh
Total Metal Impurities≤0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only