Thermo Scientific Chemicals
Stainless Steel powder, -100 mesh, Type 316-L, Thermo Scientific Chemicals
Catalog number: 011089.A3
2 kg, Each
Thermo Scientific Chemicals
Stainless Steel powder, -100 mesh, Type 316-L, Thermo Scientific Chemicals
Catalog number: 011089.A3
2 kg, Each
Quantity
Catalog number: 011089.A3
also known as 011089-A3
Price (USD)
Quantity
-
Composition
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Particle Size
-100 mesh (less than 150micron)
Description
It has an extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment. It finds its application in diamond tools, grinding and P/M parts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov