Aluminum bromide, 98%
Aluminum bromide, 98%
Aluminum bromide, 98%
Thermo Scientific Chemicals

Aluminum bromide, 98%

CAS: 7727-15-3 | AlBr3 | 266.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Catalog number 011113.09
also known as 011113-09
Price (EUR)
35,10
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
35,10
Each
Chemical Identifiers
CAS7727-15-3
IUPAC Namealuminium(3+) tribromide
Molecular FormulaAlBr3
InChI KeyPQLAYKMGZDUDLQ-UHFFFAOYSA-K
SMILES[Al+3].[Br-].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Alkylation, bromination and isomerization catalyst in organic synthesis.Aluminum bromide acts as a strong Lewis acid and is used in Lewis acid-promoted reactions such as epoxide ring openings. It is used as a catalyst for Friedel-Crafts alkylation reaction. It also finds application in bromination and isomerization reactions in organic synthesis. It is also used in polymerization reaction to prepare poly(o-xylene)from benzocyclobutene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only