Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis)
Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis)
Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis)
Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis)

CAS: 10361-46-3 | BiNO4 | 286.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011122.18
also known as 011122-18
Price (EUR)
693,00
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
693,00
Each
Add to cart
Chemical Identifiers
CAS10361-46-3
IUPAC Name(nitrooxy)bismuthanone
Molecular FormulaBiNO4
InChI KeyOGCYGKRWXWQOAK-UHFFFAOYSA-N
SMILES[O-][N+](=O)O[Bi]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Bismuth(III) nitrate oxide is used as an organic chemical synthesis intermediate. It is also used in the spectrophotometric detection of bismuth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only