Germanium pieces, 2cm (0.8in) and down, Puratronic™, 99.999% (metals basis)
Germanium pieces, 2cm (0.8in) and down, Puratronic™, 99.999% (metals basis)
Germanium pieces, 2cm (0.8in) and down, Puratronic™, 99.999% (metals basis)
Germanium pieces, 2cm (0.8in) and down, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Germanium pieces, 2cm (0.8in) and down, Puratronic™, 99.999% (metals basis)

CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 011144.22
also known as 011144-22
Price (USD)/ Each
1,245.65
Special offer
Online exclusive
1,465.00
Save 219.35 (15%)
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
1,245.65
Special offer
Online exclusive
1,465.00
Save 219.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Namegermanium
Molecular FormulaGe
InChI KeyGNPVGFCGXDBREM-UHFFFAOYSA-N
SMILES[Ge]
Molecular Weight (g/mol)72.63
View more
SpecificationsSpecification SheetSpecification Sheet
FormPieces
Total Metal Impurities<0.001%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only