Thermo Scientific Chemicals

L-(+)-Ascorbic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 011188.A3
2 kg, Each
Thermo Scientific Chemicals

L-(+)-Ascorbic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 011188.A3
2 kg, Each
Quantity
Catalog number: 011188.A3
also known as 011188-A3
Price (USD)
Price: 217.00
Online price: 187.65
Your price:
Quantity
-

Chemical Identifiers

CAS
50-81-7
IUPAC Name
(5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular Formula
C6H8O6
InChI Key
CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
Assay (Titration)
> 98.0%
Appearance (Color)
White to off-white
Optical Rotation
21.0 +/- 0.5 deg
Form
powder

Description

L-Ascorbic acid is an important nutrient and antioxidant. L-ascorbic acid is frequently used as a processing aid or as a preservative in certain nutrients or food components. It is also utilized in the synthetic polymer industry, in photo processing and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents and in assay reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text