Thermo Scientific Chemicals

Terbium(III,IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011208.06
5 g, Each
Thermo Scientific Chemicals

Terbium(III,IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011208.06
5 g, Each
Quantity
Catalog number: 011208.06
also known as 011208-06
Price (USD)
Price: 185.00
Online price: 157.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12037-01-3
IUPAC Name
tetraoxotetraterboxane
Molecular Formula
O7Tb4
InChI Key
UWZVBPNIJCKCMC-UHFFFAOYSA-N
SMILES
O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

Terbium(III,IV) oxide is used in the preparation of other terbium compounds. Activating agents for fluorescent powders, composites for catalytic automobile exhaust converters, Glass materials with a Faraday Rotation effect for optical and laser-based devices, Magneto-optical glasses, Magneto-optical recording materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text