Nitrato de escandio(III) hidrato, REacton™, 99,99% (REO), Thermo Scientific Chemicals
Nitrato de escandio(III) hidrato, REacton™, 99,99% (REO), Thermo Scientific Chemicals
Nitrato de escandio(III) hidrato, REacton™, 99,99% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitrato de escandio(III) hidrato, REacton™, 99,99% (REO), Thermo Scientific Chemicals

CAS: 107552-14-7 | N3O9Sc | 230.968 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo 011221.03
también denominado 011221-03
Precio (EUR)
164,00
Each
Cantidad:
1 g
Pedido a granel o personalizado
Precio (EUR)
164,00
Each
Identificadores químicos
CAS107552-14-7
IUPAC Namescandium(3+) trinitrate
Molecular FormulaN3O9Sc
InChI KeyDFCYEXJMCFQPPA-UHFFFAOYSA-N
SMILES[Sc+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Total Rare Earth Oxide Impurities0.01% max.
Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only