Thermo Scientific Chemicals

Samarium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011229.A1
1 kg, Each
Thermo Scientific Chemicals

Samarium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 011229.A1
1 kg, Each
Quantity
Catalog number: 011229.A1
also known as 011229-A1
Price (USD)
Price: 894.00
Online price: 774.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12060-58-1
IUPAC Name
disamarium(3+) trioxidandiide
Molecular Formula
O3Sm2
InChI Key
FKTOIHSPIPYAPE-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Sm+3].[Sm+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. It finds use as a neutron absorber in nuclear power reactors to control the reaction. It is also used as a catalyst for the dehydration of primary alcohols to aldehydes or ketones. It forms samarium perchlorate when treated with perchloric acid, which is used as a reference material for checking the wavelength scale of a spectrophotometer in the range of 200 to 500 nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text