Thermo Scientific Chemicals

Praseodymium(III, IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

100 g, Each
Thermo Scientific Chemicals

Praseodymium(III, IV) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

100 g, Each
Quantity
Catalog number: 011235.22
also known as 011235-22
Price (USD)
Price: 778.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
12037-29-5
IUPAC Name
hexapraseodymium(3+) undecaoxidandiide
Molecular Formula
O11Pr6
InChI Key
HPZIIFVSYNLWNX-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]
Total Rare Earth Oxide Impurities
0.01% max.

Description

Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only