Thermo Scientific Chemicals

Praseodymium(III) fluoride, anhydrous, REacton™, 99.9895% (REO), Thermo Scientific Chemicals

Catalog number: 011241.09
10 g, Each
Thermo Scientific Chemicals

Praseodymium(III) fluoride, anhydrous, REacton™, 99.9895% (REO), Thermo Scientific Chemicals

Catalog number: 011241.09
10 g, Each
Quantity
Catalog number: 011241.09
also known as 011241-09
Price (USD)
Price: 142.00
Online price: 123.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13709-46-1
IUPAC Name
praseodymium(3+) trifluoride
Molecular Formula
F3Pr
InChI Key
BOTHRHRVFIZTGG-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Pr+3]
Total Rare Earth Oxide Impurities
0.0105% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text