Thermo Scientific Chemicals

Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011245.06
5 g, Each
Thermo Scientific Chemicals

Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog number: 011245.06
5 g, Each
Quantity
Catalog number: 011245.06
also known as 011245-06
Price (USD)
Price: 45.50
Online price: 38.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13746-96-8
IUPAC Name
neodymium(3+) trinitrate
Molecular Formula
N3NdO9
InChI Key
CFYGEIAZMVFFDE-UHFFFAOYSA-N
SMILES
[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.01% max.

Description

It is used as pharmaceutical intermediates. It is used in the selective PVC membrane sensors. The sensor with ionophore (L1) exhibits significantly enhanced selectivity towards neodymium (III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text