Neodymium(III) fluoride, anhydrous, 99.9% (REO)
Neodymium(III) fluoride, anhydrous, 99.9% (REO)
Neodymium(III) fluoride, anhydrous, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium(III) fluoride, anhydrous, 99.9% (REO)

CAS: 13709-42-7 | F3Nd | 201.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011246.14
also known as 011246-14
Price (EUR)
154,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
154,00
Each
Chemical Identifiers
CAS13709-42-7
IUPAC Nameneodymium(3+) trifluoride
Molecular FormulaF3Nd
InChI KeyXRADHEAKQRNYQQ-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Neodymium(III) fluoride, is used extensively in the preparation and study of fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only