Nitrato de lantano(III) hexahidrato, 99,99 % (REO), Thermo Scientific Chemicals, REacton™
Nitrato de lantano(III) hexahidrato, 99,99 % (REO), Thermo Scientific Chemicals, REacton™
Nitrato de lantano(III) hexahidrato, 99,99 % (REO), Thermo Scientific Chemicals, REacton™
Thermo Scientific Chemicals

Nitrato de lantano(III) hexahidrato, 99,99 % (REO), Thermo Scientific Chemicals, REacton™

CAS: 10277-43-7 | H12LaN3O15 | 433.01 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
50 g
250 g
1 kg
Número de catálogo 011267.A1
también denominado 011267-A1
Precio (EUR)
1.027,00
Each
Cantidad:
1 kg
Pedido a granel o personalizado
Precio (EUR)
1.027,00
Each
Identificadores químicos
CAS10277-43-7
IUPAC Namelanthanum(3+) hexahydrate trinitrate
Molecular FormulaH12LaN3O15
InChI KeyGJKFIJKSBFYMQK-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Total Rare Earth Oxide Impurities0.01% max.
Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only