Cerium(IV) sulfate hydrate, REacton™, 99% (REO)
Cerium(IV) sulfate hydrate, REacton™, 99% (REO)
Cerium(IV) sulfate hydrate, REacton™, 99% (REO)
Cerium(IV) sulfate hydrate, REacton™, 99% (REO)
Thermo Scientific Chemicals

Cerium(IV) sulfate hydrate, REacton™, 99% (REO)

CAS: 95838-16-7 | CeO8S2 | 332.23 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
產品號碼 011323.22
亦稱為 011323-22
價格 (TWD)
-
申請報價
Quantity:
100 g
Request bulk or custom format
化學識別
CAS95838-16-7
IUPAC Nameλ⁴-cerium(4+) disulfate
Molecular FormulaCeO8S2
InChI KeyVZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
檢視更多
規格Specification Sheet規格表
Total Rare Earth Oxide Impurities1% max.
Cerium(IV) sulfate hydrate acts as oxidizing agent which is used for the generation of o-benzoquinone from catechol. It promotes the formation of radicals from dialkyl malonates, which undergo substitution reactions with furans and thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only