Thermo Scientific Chemicals

Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals

Catalog number: 011332.36
500 g, Each
Thermo Scientific Chemicals

Cerium(IV) ammonium nitrate, REacton™, 99.5% (REO), Thermo Scientific Chemicals

Catalog number: 011332.36
500 g, Each
Quantity
Catalog number: 011332.36
also known as 011332-36
Price (USD)
Price: 390.00
Online price: 331.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16774-21-3
IUPAC Name
λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular Formula
CeH8N8O18
InChI Key
WIBGOERAEYJBOT-UHFFFAOYSA-N
SMILES
N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Rare Earth Oxide Impurities
0.5% max.

Description

Cerium(IV) ammonium nitrate is used to offers a novel route to tartronic acid derivatives via facile oxygenation of malonates. It is also used as for oxidative addition reactions of electrophilic radicals to alkenes, enabling intermolecular and intramolecular carbon-carbon and carbon-heteroatom bond formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text