Cobalt(II) chloride hexahydrate, 99.9% (metals basis)
Cobalt(II) chloride hexahydrate, 99.9% (metals basis)
Cobalt(II) chloride hexahydrate, 99.9% (metals basis)
Cobalt(II) chloride hexahydrate, 99.9% (metals basis)
Cobalt(II) chloride hexahydrate, 99.9% (metals basis)
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, 99.9% (metals basis)

Sensitivity: Hygroscopic; m.p. 52-56°C -4H2O, 100°C -5H2O, 120-140°C -6H2O | CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 011344.30
also known as 011344-30
Price (EUR)
95,20
Online Exclusive
112,00
Save 16,80 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
95,20
Online Exclusive
112,00
Save 16,80 (15%)
Each
Add to cart
Chemical Identifiers
CAS7791-13-1
IUPAC Nameλ²-cobalt(2+) hexahydrate dichloride
Molecular FormulaCl2CoH12O6
InChI KeyGFHNAMRJFCEERV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Cobalt(II) chloride hexahydrate is used as a humidity indicator in weather instruments. It finds use in electroplating of cobalt, in invisible ink, in organic chemistry and is a precursor to cobaltocene(bis (cyclopentadienyl) cobalt(II)).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only