Thermo Scientific Chemicals

Titanium boride, Thermo Scientific Chemicals

Catalog number: 011364.30
250 g, Each
Thermo Scientific Chemicals

Titanium boride, Thermo Scientific Chemicals

Catalog number: 011364.30
250 g, Each
Quantity

Chemical Identifiers

CAS
12045-63-5
IUPAC Name
diboron titanium
Molecular Formula
B2Ti
InChI Key
QYEXBYZXHDUPRC-UHFFFAOYSA-N
SMILES
[B].[B].[Ti]
Impurity content
Oxygen 0.5% max

Description

Titanium boride is used in seals wear parts and cutting tools. It is used to make ballistic armour due to its high hardness. It is used to constitute composite materials in which the presence of the material serves to increase strength and fracture toughness of the matrix.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text