Thermo Scientific Chemicals

Niobium(V) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 011366.22
100 g, Each
Thermo Scientific Chemicals

Niobium(V) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 011366.22
100 g, Each
Quantity
Catalog number: 011366.22
also known as 011366-22
Price (USD)
Price: 135.00
Online price: 114.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1313-96-8
IUPAC Name
[(dioxoniobio)oxy]dioxoniobium
Molecular Formula
Nb2O5
InChI Key
ZKATWMILCYLAPD-UHFFFAOYSA-N
SMILES
O=[Nb](=O)O[Nb](=O)=O
Total Metal Impurities
0.1% max.

Description

Niobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in the food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators including linear and non-linear optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text