Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)

Form: approx. 0.49g/25x25mm | CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog number 011381.GH
also known as 011381-GH
Price (USD)/ Each
416.65
Special offer
Online exclusive
490.00
Save 73.35 (15%)
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
416.65
Special offer
Online exclusive
490.00
Save 73.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
Molecular Weight (g/mol)55.85
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.25mm (0.01in) thick.
Assay from Supplier's CofA99.95% (metals basis)
FormFoil, 0.1mm (0.004in) thick.
Assay from Supplier's CofA≥99.995% (metals basis)
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only