Silver powder, APS 1-3 micron, 99.9% (metals basis)
Silver powder, APS 1-3 micron, 99.9% (metals basis)
Silver powder, APS 1-3 micron, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver powder, APS 1-3 micron, 99.9% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 011405.09
also known as 011405-09
Price (USD)
81.90
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
81.90
Each
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only