Thermo Scientific Chemicals

Ammonium perrhenate(VII), 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011410.04
2 g, Each
Thermo Scientific Chemicals

Ammonium perrhenate(VII), 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 011410.04
2 g, Each
Quantity
Catalog number: 011410.04
also known as 011410-04
Price (USD)
Price: 51.90
Online price: 44.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13598-65-7
IUPAC Name
rheniumoylol amine
Molecular Formula
H4NO4Re
InChI Key
XPSXHXWHJAIQJR-UHFFFAOYSA-M
SMILES
N.O[Re](=O)(=O)=O
Total Metal Impurities
1% max.

Description

Ammonium perrhenate(VII) is used to synthesis the rehenium(III) organohydrazides which is used as radiopharmaceuticals and also used in the manufacturing of rhenium metal and its additives in superalloys. It is used as catalyst for the synthesis of platinum-rhenium catalysts which used in gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text