Thermo Scientific Chemicals

Silver sulfide, 99%, Thermo Scientific Chemicals

Catalog number: 011416.22
100 g, Each
Thermo Scientific Chemicals

Silver sulfide, 99%, Thermo Scientific Chemicals

Catalog number: 011416.22
100 g, Each
Quantity
Catalog number: 011416.22
also known as 011416-22
Price (USD)
Price: 753.00
Online price: 640.65
Your price:
Quantity
-

Chemical Identifiers

CAS
21548-73-2
IUPAC Name
disilver(1+) sulfanediide
Molecular Formula
Ag2S
InChI Key
XUARKZBEFFVFRG-UHFFFAOYSA-N
SMILES
[S--].[Ag+].[Ag+]
Appearance (Color)
Dark grey to black
Form
Powder
Assay from Suppliers CofA
≥98.5%

Description

Silver sulfide is useful as a photosensitizer in photography, Intermediates, chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text